MethElute™ Reagent (TMPAH)
MethElute™ Reagent (TMPAH)
Actual product may vary
MethElute™ Reagent (TMPAH)
For accurate, sensitive on-column methylation use Thermo Scientific™ MethElute™ Reagent (TMPAH).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
TPCTS-4930112 x 1 mL
TS-4930112 x 1 mL
Catalog number TPCTS-49301
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
12 x 1 mL
For accurate, sensitive, on-column methylation and detection by GC analysis use Thermo Scientific™ MethElute™ Reagent (TMPAH). It is ideally suited for the quantitative methylation and detection of barbiturates, sedatives, xanthine bases, phenolic alkaloids, and phenytoin, giving a single quantitative peak for each substance. When the reagent is heated with drug-containing extracts, serum, or urine the drugs containing reactive amino, hydroxyl, and carboxyl functions will be methylated at the reactive sites.
Performance Characteristics
Accuracy comparable or better than the UV/TLC method (GC is superior to UV/TLC when phenobarbitol and phenytoin present)
High precision, with coefficient of variation of 5% or less
High sensitivity; barbiturates deteced down to 0.2 mg/dL
For Research Use Only. Not for use in diagnostic procedures.
Specifications
StorageRoom Temperature
For Use WithGC
HazardsFlammable, Toxic
Standard ComponentsTrimethylanilinium hydroxide in Methanol
Standard Concentration0.2 M
Item Description12 x 1 mL
Quantity12 x 1 mL
Unit SizePack