Search Thermo Fisher Scientific
| Catalog Number | dimQuantity |
|---|---|
| SPPCRM-PE002 |
| Analyte | CAS No. | Concentration |
|---|---|---|
| Diisodecyl phthalate | 26761-40-0 | 30,000μg/g |
| Diisononyl phthalate | 28553-12-0 | 30,000μg/g |
| Bis(2-Ethylhexyl)phthalate | 117-81-7 | 3000μg/g |
| Bisphenol A | 80-05-7 | 3000μg/g |
| Butylbenzyl phthalate | 85-68-7 | 3000μg/g |
| Diethyl phthalate | 84-66-2 | 3000μg/g |
| Dimethyl phthalate | 131-11-3 | 3000μg/g |
| Di-n-butyl phthalate | 84-74-2 | 3000μg/g |
| Di-n-octyl phthalate | 117-84-0 | 3000μg/g |