Make procedures as painless as possible with extremely sharp and reliable stainless steel needles. Terumo™ Agani™ Single-use Sterile Hypodermic Needles feature a unique, latex-free double bevel design for maximum comfort.
Have Questions?
Catalog Number
Needle Gauge
Color
Needle Point Style
Length (Metric)
SMCAN2325R1
23 G
Blue
11° Regular
25 mm
Catalog number SMCAN2325R1
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Needle Gauge:
23 G
Color:
Blue
Needle Point Style:
11° Regular
Length (Metric):
25 mm
Constructed from polypropylene, stainless steel, epoxy resin and dimethyl silicone oil
All needles are supplied sterile wrapped in individual, tamper proof peel pouches
Simple color coded system allows easy identification of required sizes
Use with a wide range of Sterile Syringes
Needles are sterilized with ethylene oxide to a Sterility Assurance Level of at least 10-6 Class IIa, sterile product following MDD 93/42/EEC
CE marked and latex free
Supplied in boxes of 100 needles
Specifications
ColorBlue
Needle Gauge23 G
Diameter (Metric) Inner0.6 mm
Latex FreeYes
Length (Metric)25 mm
Product TypeSterile Hypodermic Needle for Single-Use