Savant™ DNA SpeedVac™ Concentrators Rotors
Savant™ DNA SpeedVac™ Concentrators Rotors
Actual product may vary
Savant™ DNA SpeedVac™ Concentrators Rotors
Savant™ DNA SpeedVac™ Concentrators Rotors
Savant™ DNA SpeedVac™ Concentrators Rotors
Savant™ DNA SpeedVac™ Concentrators Rotors
Thermo Fisher Scientific
Savant™ DNA SpeedVac™ Concentrators Rotors
Configure Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors with Savant DNA SpeedVac Concentrators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHolds
RD3636
RD2MP2 Shallow-well Plates
RD4848
Catalog number RD36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Holds:
36
Configure Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors with Savant DNA SpeedVac Concentrators.
Easy to use circular rotors with different tube hole sizes to accommodate different sample sizes. These rotors require no additional accessories and fit directly into your concentrator.
Specifications
Description36 x 1.5mL microtubes
TypeConcentrator Rotor
For Use With (Equipment)DNA130
Holds36
Rotor SystemTraditional
Volume (Metric)1.5 mL
Unit Size