Accessories for Wellwash™ and Wellwash™ Versa Microplate Washers
Product Image
Thermo Fisher Scientific
Accessories for Wellwash™ and Wellwash™ Versa Microplate Washers
Enhance your Wellwash and Wellwash Versa microplate washers with a range of accessories and replacement parts. Wellwash microplate washers enable reliable ELISA plate washing for 96- or 384-well plates with low residual wash volumes and user-friendly features for optimal convenience and safety.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
N10805Wash BottleWellwash Microplate Strip Washer
N10800Wash Head (1 x 8)Wellwash Microplate Strip Washer
N10801Wash Head (1 x 12)Wellwash Microplate Strip Washer
N10814Waste BottleWellwash Versa Microplate Strip Washer
Catalog number N10805
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Wash Bottle
For Use With (Equipment):
Wellwash Microplate Strip Washer
These accessories and replacement parts are for use with the Wellwash and Wellwash Versa microplate washers. Some are compatible with both models, while others are specifically designed for either the Wellwash or Wellwash Versa microplate washer.
Wellwash microplate strip washers provide reliable and robust plate washing for ELISA applications with low residual wash volumes. Wellwash microplate washers feature a large color display with graphical user interface, local language versions, and non-pressurized bottles for optimal ease of use, convenience, and safety.
Specifications
Capacity (Metric)2 L
Quantity1 Ea.
TypeWash Bottle
IncludesCap with Tubing and Liquid Level Sensors
For Use With (Equipment)Wellwash Microplate Strip Washer
Unit Size