Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Culture Area | Closure Type |
---|---|---|---|
LSVFB012935 | 50 mL | 25 cm² | Vented |
FB012935 | 50 mL | 25 cm² | Vented |
FB012937 | 250 mL | 75 cm² | Vented |
FISFB012937 | 250 mL | 75 cm² | Vented |
LSVFB012937 | 250 mL | 75 cm² | Vented |
FB012939 | 600 mL | 182 cm2 | Vented |
LSVFB012939 | 600 mL | 182 cm2 | Vented |
FISFB012941 | 850 mL | 300 cm2 | Vented |