Flat Bouffant Caps
Flat Bouffant Caps
Actual product may vary
Flat Bouffant Caps
Flat Bouffant Caps
Flat Bouffant Caps
Flat bouffant caps provide protection against dirt, grime, and particulates in non-hazardous environments. Available in blue or white in a variety of sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeColor
IENS110NW-2424 in.White
IENS110NW-24B24 in.Blue
Catalog number IENS110NW-24
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size:
24 in.
Color:
White
  • Provide a particulate barrier and helps prevent hair from contaminating a product or work environment
  • Low-linting, lightweight cover restrains hair and offers great ventilation
  • Designed to protect against dirt, grime, and particulates in non-hazardous environments
  • Flat style
  • Latex-free elastic
  • Possible applications include food processing, construction, pharmaceuticals, environmental cleanup, and more
Specifications
Product TypeBouffant
Size24 in.
MaterialPolypropylene
ColorWhite
DisposableSingle-use
ModelS110NW
Diameter (English)24 in.
For Use With (Application)Food Processing, Construction, Pharmaceuticals, Environmental Cleanup
Unit Size