TC120 Series Digital Heated Circulating Bath, Stainless Steel Tank
TC120 Series Digital Heated Circulating Bath, Stainless Steel Tank
Actual product may vary
TC120 Series Digital Heated Circulating Bath, Stainless Steel Tank
Versatile mid-range Optima heated circulator and stainless steel tank with comprehensive specification to suit most applications for precision temperature control.
Have Questions?
Catalog NumberCapacity (Metric)Dimensions (L x W x H)
GRATC120-ST3838 L730 x 333 x 450 mm
Catalog number GRATC120-ST38
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
38 L
Dimensions (L x W x H):
730 x 333 x 450 mm
  • Stability ±0.05°C.
  • Clear, bright 4 digit LED display
  • Simple, intuitive user interface: dial and two function buttons.
  • Model available with/without clamp (TClamp)
  • Low-liquid detection (float switch)
  • Fixed over temperature
  • 2 point user calibration
  • Visual alarm
  • Integral pump for external circulation
  • Visual alarm
Specifications
Capacity (Metric)38 L
Flow Rate16 L/min.
Temperature Range0°C to 120°C
External CirculationYes
MaterialStainless Steel Tank
Max. Pressure210 mbar
Dimensions (L x W x H)730 x 333 x 450 mm
Voltage120/230 V
Unit Size