Cover for General Purpose Water Bath
Product Image
Cover for General Purpose Water Bath
Fisherbrand™ Covers are for use withShallow-Form Shaking Water Baths.
Have Questions?
Catalog NumberFor Use With (Equipment)
GPSSL05Precision 2L and 5L General Purpose Water Baths
Catalog number GPSSL05
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
Precision 2L and 5L General Purpose Water Baths
Gassing Hoods
  • Permit control over the atmosphere surrounding the sample
  • Improve temperature uniformity and reduce energy consumption
  • Polished stainless steel with plastic gas fitting
  • Each bath will accommodate 1 large hood or 2 small hoods
Gable Covers
  • Enhanced hinged design supports open lid configuration and easy lid removal
  • Accommodate taller labware with extended height
  • Polycarbonate and stainless steel models available
Specifications
DescriptionStainless Steel Gable Cover
For Use With (Equipment)Precision 2L and 5L General Purpose Water Baths
TypeGable Cover