Clozapine, Thermo Scientific Chemicals
Product Image
Thermo Scientific Chemicals
Clozapine, Thermo Scientific Chemicals
CAS: 5786-21-0 | C18H19ClN4 | 326.83 g/mol
Have Questions?
Catalog NumberQuantity
FSA4642800505 g
Catalog number FSA464280050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7789-20-0
IUPAC Name[(²H)oxy](²H)
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-ZSJDYOACSA-N
SMILES[2H]O[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
D-Enrichment>=99.95 %