Gliclazide, Thermo Scientific Chemicals
Gliclazide, Thermo Scientific Chemicals
Gliclazide, Thermo Scientific Chemicals
Gliclazide, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gliclazide, Thermo Scientific Chemicals
CAS: 21187-98-4 | C15H21N3O3S | 323.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA461070010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS110-82-7
IUPAC Namecyclohexane
Molecular FormulaC6H12
InChI KeyXDTMQSROBMDMFD-UHFFFAOYSA-N
SMILESC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentFluorescence at emission maximum for Impurities: <1 ppb as Quinine sulfate
Water Content (Karl Fischer Titration)≤0.02%
Optical Absorbance at254 nm: ≤0.01 AU
Residue on Evaporation≤1ppm