Ammonium hydroxide, for HPLC, 35% solution in water
Ammonium hydroxide, for HPLC, 35% solution in water
Ammonium hydroxide, for HPLC, 35% solution in water
Ammonium hydroxide, for HPLC, 35% solution in water
Thermo Scientific Chemicals
Ammonium hydroxide, for HPLC, 35% solution in water
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Catalog NumberQuantity
FSA460801000100 mL
Catalog number FSA460801000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS7647-14-5
IUPAC Namesodium chloride
Molecular FormulaClNa
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES[Na+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 280 nm A: =<0.01
Sulfate (SO4)=<4 ppm
Iron (Fe)=<2 ppm
Calcium (Ca)=<50 ppm
Magnesium (Mg)=<50 ppm