Methyl palmitoleate, 99%, analytical standard for GC
Methyl palmitoleate, 99%, analytical standard for GC
Methyl palmitoleate, 99%, analytical standard for GC
Methyl palmitoleate, 99%, analytical standard for GC
Thermo Scientific Chemicals
Methyl palmitoleate, 99%, analytical standard for GC
CAS: 1120-25-8 | C17H32O2 | 268.44 g/mol
Have Questions?
Catalog NumberdimQuantity
FSA460400010
Catalog number FSA460400010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS34946-82-2
IUPAC Namecopper(II) triflate
Molecular FormulaC2H2CuF6O4S2
InChI KeyTXWAGEFXPDWAQH-UHFFFAOYSA-N
SMILES[Cu].FC(F)(F)S(=O)=O.FC(F)(F)S(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (special)≥98.5%
Appearance (Color)White