Trimethylsilanol, Thermo Scientific Chemicals
Trimethylsilanol, Thermo Scientific Chemicals
Trimethylsilanol, Thermo Scientific Chemicals
Trimethylsilanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trimethylsilanol, Thermo Scientific Chemicals
CAS: 1066-40-6 | C3H9LiOSi | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA458570050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS34893-92-0
IUPAC Name1,3-dichloro-5-isocyanatobenzene
Molecular FormulaC7H3Cl2NO
InChI KeyXEFUJGURFLOFAN-UHFFFAOYSA-N
SMILESClC1=CC(=CC(Cl)=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=95 %
Appearance (Color)White to light yellow
Appearance (Form)Low melting solid
Infrared spectrumConforms
Melting point29°C to 35°C