Acetamide, 99%, for analysis, Thermo Scientific Chemicals
Acetamide, 99%, for analysis, Thermo Scientific Chemicals
Acetamide, 99%, for analysis, Thermo Scientific Chemicals
Acetamide, 99%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetamide, 99%, for analysis, Thermo Scientific Chemicals
CAS: 60-35-5 | C2H5NO | 59.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number FSA451870010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetamide
CAS60-35-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Suspected of causing cancer.
Health Hazard 3GHS P Statement
Obtain special instructions before use.