3-Oxetanone, 95%, Thermo Scientific Chemicals
3-Oxetanone, 95%, Thermo Scientific Chemicals
3-Oxetanone, 95%, Thermo Scientific Chemicals
3-Oxetanone, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Oxetanone, 95%, Thermo Scientific Chemicals
CAS: 6704-31-0 | C3H4O2 | 72.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA451200010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Oxetanone
CAS6704-31-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point140°C