(-)-Epigallocatechin, 95%, from green tea
(-)-Epigallocatechin, 95%, from green tea
(-)-Epigallocatechin, 95%, from green tea
(-)-Epigallocatechin, 95%, from green tea
Thermo Scientific Chemicals
(-)-Epigallocatechin, 95%, from green tea
CAS: 970-74-1 | C15H14O7 | 306.27 g/mol
Have Questions?
Catalog NumberQuantity
FSA44898010010 mg
Catalog number FSA448980100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mg
Request bulk or custom format
Chemical Identifiers
CAS3264-82-2
IUPAC Namenickel(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14NiO4
InChI KeyBMGNSKKZFQMGDH-FDGPNNRMSA-L
SMILES[Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green
FormPowder or crystalline powder or crystals
Assay from Supplier's CofANickel, Ni : 21.5-24.2%
Assay from Supplier's CofA≥94.0 to ≤106.0% (Titration)