Carbon disulfide, 99.9%, ACS reagent
Carbon disulfide, 99.9%, ACS reagent
Carbon disulfide, 99.9%, ACS reagent
Carbon disulfide, 99.9%, ACS reagent
Thermo Scientific Chemicals
Carbon disulfide, 99.9%, ACS reagent
CAS: 75-15- | CS2 | 76.13 g/mol
Have Questions?
Catalog NumberChemical Name or Material
FSA447035000Carbon disulfide
Catalog number FSA447035000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Chemical Name or Material:
Carbon disulfide
Request bulk or custom format
Chemical Identifiers
CAS2999-46-4
IUPAC Nameethyl 2-isocyanoacetate
Molecular FormulaC5H7NO2
InChI KeyFPULFENIJDPZBX-UHFFFAOYSA-N
SMILESCCOC(=O)C[N+]#[C-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4150-1.4230 @ 20°C
FormLiquid
Appearance (Color)Clear colorless to dark brown