1-Methyl-2-pyrrolidinone, 99.5%, for analysis, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, for analysis, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, for analysis, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, for analysis, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, for analysis, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 872-50-4 | C5H9NO | 99.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number FSA445790025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS2530-83-8
IUPAC Nametrimethoxy({3-[(oxiran-2-yl)methoxy]propyl})silane
Molecular FormulaC9H20O5Si
InChI KeyBPSIOYPQMFLKFR-UHFFFAOYNA-N
SMILESCO[Si](CCCOCC1CO1)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms (non-U.S. specification)
Refractive Index1.4265-1.4315 @ 20°C
Solution TestSolubility: Passes Test (U.S. specification)
Viscosity2.9-3.2 cSt (U.S. specification)
Appearance (Color)Clear colorless
View more
Suitable for battery materials development.
RUO – Research Use Only