Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number FSA442981000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS110-71-4
Specifications
Chemical Name or MaterialEthylene glycol dimethyl ether
Melting Point-69.0°C
Health Hazard 1Danger
Health Hazard 2GHS H Statement Harmful if inhaled. May damage fertility. May damage the unborn child. Highly flammable liquid and vapor. Causes skin irritation. May form explosive peroxides.
CAS110-71-4