Propidium iodide, 95%
Propidium iodide, 95%
Propidium iodide, 95%
Propidium iodide, 95%
Thermo Scientific Chemicals
Propidium iodide, 95%
CAS: 25535-16-4 | C27H34I2N4 | 668.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
1 g
Packaging:
Glass bottle
Catalog number FSA440300250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS546-67-8
IUPAC Nameλ²-lead(2+) tetraacetate
Molecular FormulaC8H12O8Pb
InChI KeyACKFDYCQCBEDNU-UHFFFAOYSA-J
SMILES[Pb++].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Iodometric>=94 %
Stabilizerca. 4 to 5 % Acetic acid
Appearance (Color)White to pink-orange