Methyl-2-isocyanatobenzoate, 97%, Thermo Scientific Chemicals
Methyl-2-isocyanatobenzoate, 97%, Thermo Scientific Chemicals
Methyl-2-isocyanatobenzoate, 97%, Thermo Scientific Chemicals
Methyl-2-isocyanatobenzoate, 97%, Thermo Scientific Chemicals
Alfa Aesar
Methyl-2-isocyanatobenzoate, 97%, Thermo Scientific Chemicals
CAS: 1793-07-3 | C9H7NO3 | 177.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number FSA440150250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl-2-isocyanatobenzoate
CAS1793-07-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic if inhaled.
May cause allergy or asthma symptoms
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre