3-tert-Butyl-4-hydroxyanisole, 98%, Thermo Scientific Chemicals
3-tert-Butyl-4-hydroxyanisole, 98%, Thermo Scientific Chemicals
3-tert-Butyl-4-hydroxyanisole, 98%, Thermo Scientific Chemicals
3-tert-Butyl-4-hydroxyanisole, 98%, Thermo Scientific Chemicals
Alfa Aesar
3-tert-Butyl-4-hydroxyanisole, 98%, Thermo Scientific Chemicals
CAS: 121-00-6 | C11H16O2 | 180.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSA439695000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS7791-12-0
IUPAC Nameλ¹-thallanylium chloride
Molecular FormulaClTl
InChI KeyGBECUEIQVRDUKB-UHFFFAOYSA-M
SMILES[Cl-].[Tl+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.