(-)-Dibenzoyl-L-tartaric acid, 98%, Thermo Scientific Chemicals
(-)-Dibenzoyl-L-tartaric acid, 98%, Thermo Scientific Chemicals
(-)-Dibenzoyl-L-tartaric acid, 98%, Thermo Scientific Chemicals
(-)-Dibenzoyl-L-tartaric acid, 98%, Thermo Scientific Chemicals
Alfa Aesar
(-)-Dibenzoyl-L-tartaric acid, 98%, Thermo Scientific Chemicals
CAS: 2743-38-6 | C18H14O8 | 358.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA438121000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material(-)-Dibenzoyl-L-tartaric acid
CAS2743-38-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0