3-Amino-6-(trifluoromethyl)-1H-indazole, 95%, Thermo Scientific™
3-Amino-6-(trifluoromethyl)-1H-indazole, 95%, Thermo Scientific™
3-Amino-6-(trifluoromethyl)-1H-indazole, 95%, Thermo Scientific™
3-Amino-6-(trifluoromethyl)-1H-indazole, 95%, Thermo Scientific™
Alfa Aesar
3-Amino-6-(trifluoromethyl)-1H-indazole, 95%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA435810050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Amino-6-(trifluoromethyl)-1H-indazole
CAS2250-55-7
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0