Chlortetracycline hydrochloride, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals
Chlortetracycline hydrochloride, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals
Chlortetracycline hydrochloride, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals
Chlortetracycline hydrochloride, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chlortetracycline hydrochloride, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals
Chlortetracycline hydrochloride, CAS # 64-72-2, is a broad-spectrum antibiotic of the tetracycline family.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA430960050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16793-91-2
IUPAC Name1-(2-bromoethyl)-2-chlorobenzene
Molecular FormulaC8H8BrCl
InChI KeyAECBVDLERUETKG-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1CCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=94.0 %
General features
• Chlortetracycline hydrochloride is a broad-spectrum antibiotic of the tetracycline family
• Receives its name from a 7-chloro substitution on one of the rings of its parent structure, tetracycline
• A bacteriostatic that inhibits the 30S ribosomal subunit and prevents peptide formation
• Can work as an antiprotozoal and antifungal
Applications
• Chlortetracycline is suitable for use in biochemistry, microbiology, in vivo research, and cell culture
• Intended for research use only
RUO – Research Use Only
General References:
  1. Chen, C.; Ray, P.; Knowlton, K.F.; Pruden, A.; Xia, K. Effect of composting and soil type on dissipation of veterinary antibiotics in land-applied manures. Chemosphere. 2018, 196:270-279.
  2. Weinroth, M.D.; Scott, H.M.; Norby, B.; Loneragan, G.H.; Noyes, N.R.; Rovira, P.; Doster, E.; Yang, X.; Woerner, D.R.; Morley, P.S.; Belk, K.E. Effects of Ceftiofur and Chlortetracycline on the Resistomes of Feedlot Cattle. Appl Environ Microbiol. 2018, 84(13), e00610-18.