p-Toluenesulfonyl isocyanate, 96%, AcroSeal™
p-Toluenesulfonyl isocyanate, 96%, AcroSeal™
p-Toluenesulfonyl isocyanate, 96%, AcroSeal™
p-Toluenesulfonyl isocyanate, 96%, AcroSeal™
Thermo Scientific Chemicals
p-Toluenesulfonyl isocyanate, 96%, AcroSeal™
CAS: 4083-64-1 | C8H7NO3S | 197.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA430761000100 mLAcroSeal™ Glass bottle
Catalog number FSA430761000
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-18-5
IUPAC Namedodecyldimethylamine
Molecular FormulaC14H31N
InChI KeyYWFWDNVOPHGWMX-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Titration>=94 % (Tertiary amine)
GC>=94 % (Dodecyl)
Appearance (Color)Clear to slightly hazy