Resveratrol, 99%
Resveratrol, 99%
Resveratrol, 99%
Alfa Aesar
Resveratrol, 99%
Resveratrol, 3,4',5-Trihydroxystilbene, CAS # 501-36-0, is a polyphenol antioxidant found in plant foods.
Have Questions?
Catalog NumberQuantityPackaging
FSA430075000500 mgGlass bottle
Catalog number FSA430075000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialResveratrol
CAS501-36-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Harmful if swallowed.
Health Hazard 3GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
View more
General description
• Resveratrol is a phytoalexin, an endogenous antiparasitic in plants
• Resveratrol is a potent antioxidant; it is found in plant foods such as grapes, blueberries and peanuts
• Resveratrol has a possible role as an NFKb inhibitor, whereby it contributes to cell survival
Applications
• Resveratrol has anti-inflammatory effects via cyclooxygenase inhibition and may also be capable of inhibiting oncogenes
• Useful for the study of cellular redox states and aging
RUO – Research Use Only
General References:
  1. Kim, E. N.; Lim, J. H.; Kim, M. Y.; Ban, T. H.; Jang, I. A.; Yoon, H. E.; Park, C. W.; Chang, Y. S.; Choi, B. S. Resveratrol, an Nrf2 activator, ameliorates aging-related progressive renal injury. Aging (Albany NY). 2018 10(1), 83-99.
  2. Hoseini, A.; Namazi, G.; Farrokhian, A.; Reiner, Ž.; Aghadavod, E.; Bahmani, F.; Asemi, Z. The effects of resveratrol on metabolic status in patients with type 2 diabetes mellitus and coronary heart disease. Food Funct. 2019 10(9), 6042-6051.