2-Aminothiazole-5-carboxylic acid, 95%, Thermo Scientific Chemicals
2-Aminothiazole-5-carboxylic acid, 95%, Thermo Scientific Chemicals
2-Aminothiazole-5-carboxylic acid, 95%, Thermo Scientific Chemicals
2-Aminothiazole-5-carboxylic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminothiazole-5-carboxylic acid, 95%, Thermo Scientific Chemicals
CAS: 40283-46-3 | C4H4N2O2S | 144.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA429570010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS90319-52-1
IUPAC Name(4R)-4-phenyl-1,3-oxazolidin-2-one
Molecular FormulaC9H9NO2
InChI KeyQDMNNMIOWVJVLY-QMMMGPOBSA-N
SMILESO=C1N[C@@H](CO1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Melting Point (clear melt)128.0-134.0?C
Optical Rotation-55.9 ± 0.1? (C=1 in chloroform)