Propargyl bromide, 80wt.% solution in toluene, stabilized, AcroSeal™, Thermo Scientific Chemicals
Propargyl bromide, 80wt.% solution in toluene, stabilized, AcroSeal™, Thermo Scientific Chemicals
Propargyl bromide, 80wt.% solution in toluene, stabilized, AcroSeal™, Thermo Scientific Chemicals
Propargyl bromide, 80wt.% solution in toluene, stabilized, AcroSeal™, Thermo Scientific Chemicals
Propargyl bromide, 80wt.% solution in toluene, stabilized, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Propargyl bromide, 80wt.% solution in toluene, stabilized, AcroSeal™, Thermo Scientific Chemicals
CAS: 106-96-7 | C3H3Br | 118.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA429121000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White-silver
FormRod, 1.27cm (0.5in) diameter.
Assay from Supplier's CofA>99.99% (metals basis)
Alkylation reagent
RUO – Research Use Only