N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™
N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™
N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™
N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™
N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™
Thermo Scientific Chemicals
N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™
CAS: 538-75-0 | C13H22N2 | 206.33 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA429021000100 mLAcroSeal™ Glass Bottle
Catalog number FSA429021000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS611-09-6
IUPAC Name5-nitro-2,3-dihydro-1H-indole-2,3-dione
Molecular FormulaC8H4N2O4
InChI KeyUNMYHYODJHKLOC-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C2NC(=O)C(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange-brown
Assay (unspecified)(Based on Carbon Elemental): >97.5 to <102.5%
FormPowder
Elemental AnalysisC: 48.76-51.26% (Theory: 50.01%)
Elemental AnalysisN: 14.17-14.99% (Theory: 14.58%)
Coupling reagent
RUO – Research Use Only