Trimethyltin Chloride, 1M (25 wt.%) solution in hexanes, AcroSeal™
Trimethyltin Chloride, 1M (25 wt.%) solution in hexanes, AcroSeal™
Trimethyltin Chloride, 1M (25 wt.%) solution in hexanes, AcroSeal™
Trimethyltin Chloride, 1M (25 wt.%) solution in hexanes, AcroSeal™
Thermo Scientific Chemicals
Trimethyltin Chloride, 1M (25 wt.%) solution in hexanes, AcroSeal™
CAS: 1066-45-1 | C3H11ClSn | 201.28 g/mol
Have Questions?
Catalog NumberQuantity
FSA427861000100 mL
Catalog number FSA427861000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS3385-94-2
IUPAC Nametrimethyl[(trimethylsilyl)sulfanyl]silane
Molecular FormulaC6H18SSi2
InChI KeyRLECCBFNWDXKPK-UHFFFAOYSA-N
SMILESC[Si](C)(C)S[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
Transition TemperatureLiquid
Assay (GC)≥97.5%