Isophorone diisocyanate, 98+%, mixture of isomers, Thermo Scientific Chemicals
Isophorone diisocyanate, 98+%, mixture of isomers, Thermo Scientific Chemicals
Isophorone diisocyanate, 98+%, mixture of isomers, Thermo Scientific Chemicals
Isophorone diisocyanate, 98+%, mixture of isomers, Thermo Scientific Chemicals
Isophorone diisocyanate, 98+%, mixture of isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isophorone diisocyanate, 98+%, mixture of isomers, Thermo Scientific Chemicals
CAS: 4098-71-9 | C12H18N2O2 | 222.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
50 mL
250 mL
Packaging:
Glass bottle
Catalog number FSA427600010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS56-89-3
IUPAC Name(2R)-2-amino-3-{[(2R)-2-amino-2-carboxyethyl]disulfanyl}propanoic acid
Molecular FormulaC6H12N2O4S2
InChI KeyLEVWYRKDKASIDU-IMJSIDKUSA-N
SMILESN[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥95.0%
Infrared spectrumConforms
Appearance (Color)White
CommentMicrobial Count: ≤1000 cfu/g (total plate count)
Loss on Drying≤0.2%