5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
CAS: 5965-83-3 | C9H14O6S | 250.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA424515000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS77301-42-9
IUPAC Name(1R)-1-(6-methoxynaphthalen-2-yl)ethan-1-ol
Molecular FormulaC13H14O2
InChI KeyOUVJWFRUESFCCY-SECBINFHSA-N
SMILESCOC1=CC=C2C=C(C=CC2=C1)[C@@H](C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light beige
HPLC>=97.5 %
Appearance (Form)Crystalline powder
Melting point111°C to 114°C
Infrared spectrumConforms
Indicator
RUO – Research Use Only