Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals
Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals
Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals
Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals
CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
View all
Glass Bottle
Catalog number FSA424040025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS62637-93-8
IUPAC NameN,N-dimethylmethanamine oxide dihydrate
Molecular FormulaC3H13NO3
InChI KeyPGFPZGKEDZGJQZ-UHFFFAOYSA-N
SMILESO.O.C[N+](C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)93.0-101.0°C
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%