Iodic acid, 99.6%, Thermo Scientific Chemicals
Iodic acid, 99.6%, Thermo Scientific Chemicals
Iodic acid, 99.6%, Thermo Scientific Chemicals
Iodic acid, 99.6%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodic acid, 99.6%, Thermo Scientific Chemicals
CAS: 7782-68-5 | ILiO3 | 181.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA423815000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20781-20-8
IUPAC Name1-(2,4-dimethoxyphenyl)methanamine
Molecular FormulaC9H13NO2
InChI KeyQOWBXWFYRXSBAS-UHFFFAOYSA-N
SMILESCOC1=CC=C(CN)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5470-1.5510 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Oxidation reagent
RUO – Research Use Only