N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
View all
Glass Bottle
Catalog number FSA423645000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS25535-16-4
IUPAC Name3,8-diamino-5-{3-[diethyl(methyl)azaniumyl]propyl}-6-phenylphenanthridin-5-ium diiodide
Molecular FormulaC27H34I2N4
InChI KeyXJMOSONTPMZWPB-UHFFFAOYSA-M
SMILES[I-].[I-].CC[N+](C)(CC)CCC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Red
Titration Argentometric94 to 106 %
Appearance (Form)Powder
Water=<5 %