Cupferron, Thermo Scientific Chemicals
Cupferron, Thermo Scientific Chemicals
Cupferron, Thermo Scientific Chemicals
Cupferron, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cupferron, Thermo Scientific Chemicals
CAS: 135-20-6 | C6H9N3O2 | 155.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA423600250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS288-36-8
IUPAC Name2H-1,2,3-triazole
Molecular FormulaC2H3N3
InChI KeyQWENRTYMTSOGBR-UHFFFAOYSA-N
SMILESN1N=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4940 to 1.4980 (20°C, 589 nm)
Infrared spectrumConforms
Titration with HClO496.0 to 104.0 %
Appearance (Color)Clear colorless to yellow