Cobalt(II) nitrate hexahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Cobalt(II) nitrate hexahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Cobalt(II) nitrate hexahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Cobalt(II) nitrate hexahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cobalt(II) nitrate hexahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
CAS: 10026-22-9 | CoH12N2O12 | 291.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
View all
Plastic bottle
Catalog number FSA423581000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS5271-38-5
IUPAC Name2-(methylsulfanyl)ethan-1-ol
Molecular FormulaC3H8OS
InChI KeyWBBPRCNXBQTYLF-UHFFFAOYSA-N
SMILESCSCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4934 to 1.4954 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=98.5 %
Appearance (Form)Liquid