Tributyl citrate, 99+%, Thermo Scientific Chemicals
Tributyl citrate, 99+%, Thermo Scientific Chemicals
Tributyl citrate, 99+%, Thermo Scientific Chemicals
Tributyl citrate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tributyl citrate, 99+%, Thermo Scientific Chemicals
CAS: 77-94-1 | C18H32O7 | 360.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA421442500
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS23142-01-0
IUPAC Name2-[2-(diethylamino)ethoxy]ethyl 1-phenylcyclopentane-1-carboxylate; 2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular FormulaC26H39NO10
InChI KeyAKJDEXBCRLOVTH-UHFFFAOYSA-N
SMILESOC(=O)CC(O)(CC(O)=O)C(O)=O.CCN(CC)CCOCCOC(=O)C1(CCCC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)95.0% min.