Thymolphthalein, ACS reagent
Thymolphthalein, ACS reagent
Thymolphthalein, ACS reagent
Thymolphthalein, ACS reagent
Thermo Scientific Chemicals
Thymolphthalein, ACS reagent
CAS: 125-20-2 | C28H30O4 | 430.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA420950250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS91-10-1
IUPAC Name2,6-dimethoxyphenol
Molecular FormulaC8H10O3
InChI KeyKLIDCXVFHGNTTM-UHFFFAOYSA-N
SMILESCOC1=CC=CC(OC)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or pale yellow to grey to brown
FormCrystalline powder or crystals or fused solid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Solution Test5% w/v in DMF: clear
View more
Indicator
RUO – Research Use Only