Tetramethylammonium perchlorate, 98%, Thermo Scientific Chemicals
Tetramethylammonium perchlorate, 98%, Thermo Scientific Chemicals
Tetramethylammonium perchlorate, 98%, Thermo Scientific Chemicals
Tetramethylammonium perchlorate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetramethylammonium perchlorate, 98%, Thermo Scientific Chemicals
CAS: 2537-36-2 | C4H12ClNO4 | 173.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA420561000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS6132-02-1
IUPAC Namedisodium decahydrate carbonate
Molecular FormulaCH20Na2O13
InChI KeyXYQRXRFVKUPBQN-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Titration with HCl>=99.0 %
Heavy metals (as Pb)=<10 ppm
Total nitrogen (as N)=<0.0005 %
PO4 and SiO2 (as SiO2)=<0.002 %
View more
Oxidation reagent, Phase transfer catalyst
RUO – Research Use Only