L(+)-Tartaric acid, ACS reagent, Thermo Scientific Chemicals
L(+)-Tartaric acid, ACS reagent, Thermo Scientific Chemicals
L(+)-Tartaric acid, ACS reagent, Thermo Scientific Chemicals
L(+)-Tartaric acid, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(+)-Tartaric acid, ACS reagent, Thermo Scientific Chemicals
CAS: 87-69-4 | C4H6O6 | 150.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA420005000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS9002-18-0
IUPAC Name2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
Molecular FormulaC14H24O9
InChI KeyGYYDPBCUIJTIBM-UHFFFAOYNA-N
SMILESCOC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤ 20.0%
AshTotal Ash: ≤ 6.5%
CommentIdentity: Passes test
FormLight tan granular powder