Orcinol, 98%, Thermo Scientific Chemicals
Orcinol, 98%, Thermo Scientific Chemicals
Orcinol, 98%, Thermo Scientific Chemicals
Orcinol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Orcinol, 98%, Thermo Scientific Chemicals
CAS: 504-15-4 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA416581000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6476-37-5
IUPAC Namedicyclohexyl(phenyl)phosphane
Molecular FormulaC18H27P
InChI KeyVPLLTGLLUHLIHA-UHFFFAOYSA-N
SMILESC1CCC(CC1)P(C1CCCCC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Formcrystalline powder
Phosphorus-31 NMRConforms to structure
Assay (GC)>97.5%