Nitro Blue Tetrazolium Chloride, 90%
Nitro Blue Tetrazolium Chloride, 90%
Nitro Blue Tetrazolium Chloride, 90%
Nitro Blue Tetrazolium Chloride, 90%
Thermo Scientific Chemicals
Nitro Blue Tetrazolium Chloride, 90%
CAS: 298-83-9 | C40H30Cl2N10O6 | 817.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
Packaging:
Glass bottle
Catalog number FSA415925000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS19455-23-3
IUPAC Namepotassium 2,2-dimethylpropanoate
Molecular FormulaC5H9KO2
InChI KeyWFMNHCSATCWAAQ-UHFFFAOYSA-M
SMILES[K+].CC(C)(C)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥94.0%
FormPowder or crystals or crystalline powder