L-alpha-Lecithin, granular, from soybean oil
L-alpha-Lecithin, granular, from soybean oil
L-alpha-Lecithin, granular, from soybean oil
L-alpha-Lecithin, granular, from soybean oil
Thermo Scientific Chemicals
L-alpha-Lecithin, granular, from soybean oil
L-alpha-Lecithin, >-97% (phospholipids) min. (as acetone insoluble), C42H80NO8P, CAS Number-8002-43-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA413102500
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS75-31-0
IUPAC Namepropan-2-amine
Molecular FormulaC3H9N
InChI KeyJJWLVOIRVHMVIS-UHFFFAOYSA-N
SMILESCC(C)N
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.3725-1.3755 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥99.0%