(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific Chemicals
(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific Chemicals
(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific Chemicals
(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific Chemicals
Alfa Aesar
(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific Chemicals
CAS: 36567-72-3 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA412120050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS122-57-6
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid or crystalline chunks or clear liquid as melt
Identification (FTIR)Conforms
Appearance (Color)Pale yellow to yellow
Assay (GC)≥98.0%