Hexyl isocyanate, 99%, Thermo Scientific Chemicals
Hexyl isocyanate, 99%, Thermo Scientific Chemicals
Hexyl isocyanate, 99%, Thermo Scientific Chemicals
Hexyl isocyanate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hexyl isocyanate, 99%, Thermo Scientific Chemicals
CAS: 2525-62-4 | C7H13NO | 127.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA411661000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS100-21-0
IUPAC Namebenzene-1,4-dicarboxylic acid
Molecular FormulaC8H6O4
InChI KeyKKEYFWRCBNTPAC-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥98.0%
Identification (FTIR)Conforms
Appearance (Color)White
FormPowder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%