Gibberellic acid, 99%, Thermo Scientific Chemicals
Gibberellic acid, 99%, Thermo Scientific Chemicals
Gibberellic acid, 99%, Thermo Scientific Chemicals
Gibberellic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gibberellic acid, 99%, Thermo Scientific Chemicals
CAS: 77-06-5 | C19H22O6 | 346.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA410910010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1120-28-1
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Free acid=<1 %
Appearance (Color)White to almost white
Appearance (Form)Powder or flakes