Gallic Acid 98%, Thermo Scientific Chemicals
Gallic Acid 98%, Thermo Scientific Chemicals
Gallic Acid 98%, Thermo Scientific Chemicals
Gallic Acid 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gallic Acid 98%, Thermo Scientific Chemicals
CAS: 149-91-7 | C7H6O5 | 170.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number FSA410862500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS50-70-4
IUPAC Namehexane-1,2,3,4,5,6-hexol
Molecular FormulaC6H14O6
InChI KeyFBPFZTCFMRRESA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Sulfated ash=<0.1 %
Appearance (Color)White to almost white
Infrared spectrumConforms
Specific optical rotation+4.0° to +7.0° (20°C, 589 nm) (c=10, borate soln.)